Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338319.1 | NP_001327296.1 | 8 - 9 | 1700 | 98 | |
NM_001338320.1 | NP_001327298.1 | 8 - 9 | 1700 | 98 | ||
NM_001338321.1 | NP_001327297.1 | 8 - 9 | 1620 | 98 | ||
NM_112686.4 | NP_566595.1 | 8 - 9 | 1220 | 98 | ||
NM_202602.2 | NP_974331.1 | 7 - 8 | 1743 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |