Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112733.3 | NP_566611.1 | 4 - 5 | 769 | 100 | |
GenBank mRNA | AY056225.1 | - | 4 - 5 | 713 | 100 | |
AY142680.1 | - | 4 - 5 | 537 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |