Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112759.5 | NP_188503.2 | 9 - 10 | 2667 | 90 | |
GenBank mRNA | AB112744.1 | - | 9 - 10 | 2625 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |