Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035646.2 | NP_001030723.1 | 4 - 5 | 298 | 121 | |
NM_001202999.2 | NP_001189928.1 | 3 - 4 | 300 | 121 | ||
NM_001338354.1 | NP_001319584.1 | 3 - 4 | 473 | 121 | ||
NM_001338356.1 | NP_001326380.1 | 3 - 4 | 157 | 121 | ||
NM_112761.3 | NP_188505.1 | 3 - 4 | 234 | 121 | ||
GenBank mRNA | AB084270.1 | - | 2 - 3 | 115 | 121 | |
AK175997.1 | - | 3 - 4 | 234 | 121 | ||
BT030341.1 | - | 2 - 3 | 115 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |