Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112791.6 | NP_188535.4 | 4 - 5 | 1019 | 93 | |
GenBank mRNA | DQ399530.1 | - | 3 - 4 | 819 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |