Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035648.1 | NP_001030725.1 | 7 - 8 | 1380 | 132 | |
NM_112805.5 | NP_188549.2 | 7 - 8 | 1419 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |