Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203004.1 | NP_001189933.1 | 2 - 3 | 671 | 78 | |
NM_001338387.1 | NP_001325757.1 | 2 - 3 | 646 | 78 | ||
NM_112808.4 | NP_188552.3 | 2 - 3 | 646 | 78 | ||
GenBank mRNA | AB250666.1 | - | 2 - 3 | 621 | 78 | |
DQ912973.1 | - | 2 - 3 | 559 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |