Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203004.1 | NP_001189933.1 | 7 - 8 | 1264 | 67 | |
NM_001338387.1 | NP_001325757.1 | 7 - 8 | 1239 | 67 | ||
NM_112808.4 | NP_188552.3 | 7 - 8 | 1239 | 67 | ||
GenBank mRNA | AB250666.1 | - | 7 - 8 | 1214 | 67 | |
DQ912973.1 | - | 7 - 8 | 1152 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |