Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112813.2 | NP_188557.1 | 4 - 5 | 1072 | 98 | |
GenBank mRNA | AK118029.1 | - | 4 - 5 | 924 | 98 | |
AY084980.1 | - | 4 - 5 | 954 | 98 | ||
BT008341.1 | - | 4 - 5 | 691 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |