Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112815.4 | NP_188559.1 | 2 - 3 | 745 | 107 | |
GenBank mRNA | AJ345084.1 | - | 2 - 3 | 609 | 107 | |
AJ404860.1 | - | 2 - 3 | 517 | 107 | ||
KJ676984.1 | - | 2 - 3 | 517 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |