Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112861.4 | NP_188605.1 | 3 - 4 | 544 | 92 | |
GenBank mRNA | AF446892.1 | - | 3 - 4 | 391 | 92 | |
AJ271732.1 | - | 3 - 4 | 440 | 92 | ||
AK226525.1 | - | 3 - 4 | 460 | 92 | ||
AY052676.1 | - | 3 - 4 | 447 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |