Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCAT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112861.4 | NP_188605.1 | 4 - 5 | 672 | 87 | |
GenBank mRNA | AF446892.1 | - | 4 - 5 | 519 | 87 | |
AJ271732.1 | - | 4 - 5 | 568 | 87 | ||
AK226525.1 | - | 4 - 5 | 588 | 87 | ||
AY052676.1 | - | 4 - 5 | 575 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |