Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112933.3 | NP_566661.1 | 3 - 4 | 1120 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |