Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112947.4 | NP_188691.1 | 7 - 8 | 924 | 80 | |
GenBank mRNA | AY062626.1 | - | 7 - 8 | 841 | 80 | |
BT000005.1 | - | 6 - 7 | 741 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |