Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112947.4 | NP_188691.1 | 9 - 10 | 1014 | 128 | |
GenBank mRNA | AY062626.1 | - | 9 - 10 | 931 | 128 | |
BT000005.1 | - | 8 - 9 | 831 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |