Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCB2;2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342363.1 | NP_001329476.1 | 2 - 3 | 348 | 97 | |
NM_119727.4 | NP_195287.1 | 2 - 3 | 348 | 97 | ||
GenBank mRNA | AK226721.1 | - | 2 - 3 | 330 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |