Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APX5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119763.3 | NP_195321.1 | 8 - 9 | 725 | 134 | |
GenBank mRNA | AK119023.1 | - | 8 - 9 | 705 | 134 | |
BT006053.1 | - | 8 - 9 | 660 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |