Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119797.4 | NP_568000.1 | 7 - 8 | 846 | 92 | |
GenBank mRNA | BT001940.1 | - | 7 - 8 | 672 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |