Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342408.1 | NP_001328464.1 | 3 - 4 | 1027 | 103 | |
NM_119801.4 | NP_568002.1 | 3 - 4 | 1089 | 103 | ||
GenBank mRNA | BT029220.1 | - | 3 - 4 | 784 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |