Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119801.4 | NP_568002.1 | 6 - 7 | 1527 | 130 | |
GenBank mRNA | BT029220.1 | - | 6 - 7 | 1222 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |