Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342408.1 | NP_001328464.1 | 7 - 8 | 1773 | 72 | |
NM_119801.4 | NP_568002.1 | 8 - 9 | 1763 | 72 | ||
GenBank mRNA | BT029220.1 | - | 8 - 9 | 1458 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |