Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036719.2 | NP_001031796.1 | 10 - 11 | 1216 | 72 | |
NM_119811.3 | NP_568005.1 | 10 - 11 | 1486 | 72 | ||
GenBank mRNA | AB063254.1 | - | 9 - 10 | 1028 | 72 | |
AY120759.1 | - | 10 - 11 | 1304 | 72 | ||
BT000131.1 | - | 9 - 10 | 1007 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |