Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SFH12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119812.4 | NP_568006.1 | 9 - 10 | 1294 | 83 | |
GenBank mRNA | AY050419.1 | - | 9 - 10 | 1188 | 83 | |
BT000834.1 | - | 9 - 10 | 1080 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |