Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119857.3 | NP_568010.1 | 2 - 3 | 700 | 123 | |
GenBank mRNA | AF319540.1 | - | 2 - 3 | 638 | 123 | |
AK229267.1 | - | 2 - 3 | 656 | 123 | ||
BT026462.1 | - | 2 - 3 | 616 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |