Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036721.2 | NP_001031798.1 | 3 - 4 | 351 | 133 | |
NM_119864.5 | NP_195418.1 | 2 - 3 | 259 | 133 | ||
GenBank mRNA | AY363869.1 | - | 2 - 3 | 153 | 133 | |
BT024826.1 | - | 2 - 3 | 153 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |