Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119867.3 | NP_568014.1 | 4 - 5 | 640 | 142 | |
GenBank mRNA | AF250963.1 | - | 4 - 5 | 601 | 142 | |
AK118314.1 | - | 4 - 5 | 592 | 142 | ||
AY085839.1 | - | 4 - 5 | 598 | 142 | ||
BT005680.1 | - | 4 - 5 | 565 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |