Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAP1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119867.3 | NP_568014.1 | 6 - 7 | 756 | 84 | |
GenBank mRNA | AF250963.1 | - | 6 - 7 | 717 | 84 | |
AK118314.1 | - | 6 - 7 | 708 | 84 | ||
AY085839.1 | - | 6 - 7 | 714 | 84 | ||
BT005680.1 | - | 6 - 7 | 681 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |