Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342434.1 | NP_001329607.1 | 3 - 4 | 537 | 62 | |
NM_119879.3 | NP_195433.1 | 3 - 4 | 689 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |