Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sks15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119879.3 | NP_195433.1 | 6 - 7 | 1495 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |