Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP81D8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119900.3 | NP_195453.1 | 1 - 2 | 560 | 123 | |
GenBank mRNA | AY065192.1 | - | 1 - 2 | 521 | 123 | |
BT000090.1 | - | 1 - 2 | 495 | 123 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |