Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRFR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342443.1 | NP_001329700.1 | 11 - 12 | 1969 | 91 | |
NM_119910.4 | NP_195462.3 | 11 - 12 | 1969 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |