Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HKL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119945.2 | NP_195497.1 | 2 - 3 | 425 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |