Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELI3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036728.2 | NP_001031805.1 | 1 - 2 | 316 | 75 | |
NM_119959.4 | NP_195511.1 | 1 - 2 | 316 | 75 | ||
GenBank mRNA | AF360225.1 | - | 1 - 2 | 136 | 75 | |
AK317050.1 | - | 1 - 2 | 136 | 75 | ||
AY040066.1 | - | 1 - 2 | 87 | 75 | ||
AY050407.1 | - | 1 - 2 | 168 | 75 | ||
AY050931.1 | - | 1 - 2 | 136 | 75 | ||
AY056385.1 | - | 1 - 2 | 166 | 75 | ||
AY302079.1 | - | 1 - 2 | 87 | 75 | ||
BT002729.1 | - | 1 - 2 | 87 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |