Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXPA20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119982.2 | NP_195534.1 | 1 - 2 | 293 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |