Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179887.3 | NP_850218.1 | 7 - 8 | 3401 | 72 | |
GenBank mRNA | AY074292.1 | - | 7 - 8 | 3328 | 72 | |
AY091326.1 | - | 5 - 6 | 2176 | 72 | ||
DQ027037.1 | - | 5 - 6 | 2176 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |