Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: URED | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179908.3 | NP_850239.1 | 3 - 4 | 246 | 86 | |
GenBank mRNA | AJ312202.1 | - | 3 - 4 | 246 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |