Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HB22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179935.2 | NP_850266.1 | 1 - 2 | 207 | 87 | |
GenBank mRNA | DQ056569.1 | - | 1 - 2 | 91 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |