Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179940.6 | NP_850271.5 | 44 - 45 | 5075 | 100 | |
GenBank mRNA | GQ373182.1 | - | 44 - 45 | 4777 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |