Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_179940.6 | NP_850271.5 | 46 - 47 | 5319 | 103 | |
GenBank mRNA | GQ373182.1 | - | 46 - 47 | 5021 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |