Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HTA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336738.1 | NP_001318377.1 | 2 - 3 | 222 | 100 | |
NM_129438.2 | NP_181415.1 | 2 - 3 | 235 | 100 | ||
NM_179967.2 | NP_850298.1 | 2 - 3 | 138 | 100 | ||
NM_179968.3 | NP_850299.1 | 1 - 2 | 361 | 100 | ||
GenBank mRNA | AK220868.1 | - | 2 - 3 | 138 | 100 | |
AK228272.1 | - | 2 - 3 | 138 | 100 | ||
AY088573.1 | - | 2 - 3 | 235 | 100 | ||
BT026423.1 | - | 1 - 2 | 117 | 100 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Arabidopsis thaliana Probe spans exons |