Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336762.1 | NP_001325386.1 | 3 - 4 | 572 | 67 | |
NM_179982.3 | NP_850313.2 | 3 - 4 | 675 | 67 | ||
GenBank mRNA | AY576276.1 | - | 3 - 4 | 430 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |