Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129531.3 | NP_565913.1 | 5 - 6 | 1375 | 77 | |
NM_179989.3 | NP_850320.1 | 5 - 6 | 1375 | 77 | ||
NM_179990.1 | NP_850321.1 | 5 - 6 | 1202 | 77 | ||
GenBank mRNA | AF325049.2 | - | 5 - 6 | 1116 | 77 | |
AY056108.1 | - | 5 - 6 | 1116 | 77 | ||
AY088487.1 | - | 5 - 6 | 1231 | 77 | ||
BT000710.1 | - | 5 - 6 | 1113 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |