Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197956.2 | NP_001184885.1 | 8 - 9 | 2074 | 63 | |
NM_100016.3 | NP_563625.1 | 6 - 7 | 1993 | 63 | ||
GenBank mRNA | AF272002.3 | - | 6 - 7 | 1993 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |