Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100028.3 | NP_171653.2 | 8 - 9 | 1165 | 124 | |
GenBank mRNA | BT028973.1 | - | 7 - 8 | 844 | 124 | |
DQ056443.1 | - | 7 - 8 | 844 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |