Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LEA14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100029.6 | NP_171654.1 | 1 - 2 | 326 | 79 | |
GenBank mRNA | AY074855.1 | - | 1 - 2 | 264 | 79 | |
BT015111.1 | - | 1 - 2 | 204 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |