Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FRO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100041.2 | NP_171665.1 | 1 - 2 | 259 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |