Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEX11C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100065.5 | NP_563636.1 | 7 - 8 | 1017 | 91 | |
GenBank mRNA | AB022861.1 | - | 7 - 8 | 739 | 91 | |
AY064139.1 | - | 7 - 8 | 792 | 91 | ||
AY086844.1 | - | 7 - 8 | 792 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |