Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197959.1 | NP_001184888.1 | 11 - 12 | 1038 | 69 | |
NM_001331318.1 | NP_001320381.1 | 11 - 12 | 1207 | 69 | ||
NM_001331319.1 | NP_001320383.1 | 10 - 11 | 1278 | 69 | ||
NM_001331320.1 | NP_001320382.1 | 10 - 11 | 1272 | 69 | ||
NM_001331321.1 | NP_001320384.1 | 5 - 6 | 566 | 69 | ||
NM_100081.5 | NP_563643.4 | 11 - 12 | 1399 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |