Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331325.1 | NP_001321166.1 | 6 - 7 | 649 | 71 | |
NM_001331326.1 | NP_001321165.1 | 6 - 7 | 649 | 71 | ||
NM_100092.3 | NP_171714.2 | 6 - 7 | 649 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |