Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331325.1 | NP_001321166.1 | 8 - 9 | 880 | 126 | |
NM_001331326.1 | NP_001321165.1 | 8 - 9 | 880 | 126 | ||
NM_100092.3 | NP_171714.2 | 8 - 9 | 880 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |